missing translation for 'onlineSavingsMsg'
Learn More

Technical Service Consultants™ Water Sampling Container

Product Code. 15733657
Change view
Click to view available options
Includes:
60 mg Sodium Thiosulphate
9 mg Sodium Thiosulphate
90 mg Sodium Thiosulphate
Unit Size:
Pack of 40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
15733657 9 mg Sodium Thiosulphate Pack of 40
15713657 60 mg Sodium Thiosulphate -
15723657 90 mg Sodium Thiosulphate -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15733657 Supplier Technical Service Consultants™ Supplier No. TS/15DL40

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sterile container with 500mL capactiy containing Sodium Thiosulphate for chlorine removal

  • 500mL Sterile Container + Sodium Thiosulphate for chlorine removal
  • Please Note: the batch & expiry details are stated on the side of the label
  • Shelf life of 2 years
  • Chlorine level must be assessed before sampling to ensure neutralisation

Specifications

Material HDPE
Description 9 mg (18 mg/liter) Sodium Thiosulphate for chlorine removal
Includes 9 mg Sodium Thiosulphate
Capacity (Metric) 500 mL
Color Clear Bottle, Blue Cap
For Use With (Application) Neutralizing 5 mg/L Chlorine per liter of water. Ideal for potable water sampling.
Quantity 40
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.