missing translation for 'onlineSavingsMsg'
Learn More

Water, TraceSELECT™, For Trace Analysis, Solstice

Product Code. 15607550
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
10 L
4 x 2.5 L
Packaging:
Cubitainer
Glass Bottle
Poly Bottle
Unit Size:
1 L
10 L
2.5 L
250mL
500mL
Pack of 4
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15607550 1 L Glass Bottle 1 L
15627550 250 mL Poly Bottle 250mL
15647550 500 mL Poly Bottle 500mL
15617550 2.5 L Poly Bottle 2.5 L
15697540 10 L Cubitainer 10 L
15637550 4 x 2.5 L Poly Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15607550 Supplier Honeywell-Riedel-de Haen Supplier No. 953051L
  • ONLINE EXCLUSIVE PRICE
    63.12 €
    49.85 € / 1 L
    Save 13.27 €
    21% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TraceSELECT™, for trace analysis

  • High purity solvents for speciation analysis by LC-ICP-MS

  • Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance

  • The blank values for metal traces in these solvents are in the ppb range or lower.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 6 to 8 (at 20°C)
Color Colorless
Melting Point 0°C
Boiling Point 100°C
UN Number NONH for all modes of transport
Beilstein 2050024
Formula Weight 18.02g/mol
Grade TraceSELECT
Odor Odorless
Packaging Glass Bottle
Density 1g/mL at 20°C
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.