missing translation for 'onlineSavingsMsg'
Learn More

Water, for UHPLC Gradient Grade Analysis, Fisher Chemical™

Product Code. 11357090 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357090 2.5 L 2.5 L
11307090 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357090 Supplier Fisher Chemical Supplier No. W/0120/PB17
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Efficiency: lot-to-lot consistency ensures reproducible results
  • Low acidity/alkalinity levels avoid interactions with samples
  • Peak shape: low baseline noise at 210nm and 254nm improves peak shape
  • Speed: filtered to 0.1μm, allowing analysis to be performed faster at higher pressures with less risk of pump blockages
  • Sensitivity: interference-free with outstandingly high UV transmission for improved resolution and sensitivity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number 11332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifica

Color Colorless
Boiling Point 100°C
Formula Weight 18.02g/mol
Packaging Plastic Coated Glass Bottle
Vapor Pressure 17.5mmHg at 20°C
Quantity 2.5 L
Physical Form Liquid
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.