missing translation for 'onlineSavingsMsg'
Learn More

Water (Ultra Pure) For Laboratory Use, AAS and ICP, Chem Lab

Product Code. 11988484
Change view
Click to view available options
Quantity:
20 L
Unit Size:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11988484 20 L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11988484 Supplier Chem-Lab Supplier No. CL02.2101.9520
  • ONLINE EXCLUSIVE PRICE
    78.70 €
    40.30 € / 20 L
    Save 38.40 €
    48% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

pH 5 to 7.5
CAS 7732-18-5
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 18.02
Packaging Ecopack (bag in box)
Density 1 g/mL
Quantity 20 L
Physical Form Liquid
Purification Method ASTM D-94, D-129
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.