missing translation for 'onlineSavingsMsg'
Learn More

Water, ULTREX™ II Ultrapure Reagent, J.T. Baker™

Product Code. 15518384 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Poly Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15518384 1 L Narrow Mouth Poly Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15518384 Supplier Avantor J.T.Baker Supplier No. 690602

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagents for critical elemental analysis with less than 10 parts-per-trillion (ppt) levels of up to 65 elements

    For use in ICP (Inducutively Coupled Plasma Spectrometry), Trace Metal Analysis

Specifications

CAS 7732-18-5
Molecular Formula H2O
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
PubChem CID 962
ChEBI CHEBI:15377
Formula Weight 18
Grade ULTREX™ II Ultrapure Reagent
Packaging Narrow Mouth Poly Bottle
Quantity 1 L
Specific Gravity 1L = 1.00kg
Chemical Name or Material Water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.