missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Waterproof Portable pH Meter

Product Code. 10713931
Change view
Click to view available options
Dimensions (L x W x H):
185 x 72 x 36 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
10713931 185 x 72 x 36 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10713931 Supplier Hanna Instruments™ Supplier No. HI9124

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Accuracy (Temperature) ±0.4°C
Accuracy (pH) ±0.01
Calibration Automatic/Manual
Range (pH) -2 to 16
Resolution (pH) 0.01
Calibration Points 1 or 2 with 5 memorized buffers (4.01, 6.86, 7.01, 9.18 and 10.01 pH)
Temperature Range (Metric) 20°C to 120°C
Resolution (Temperature) 0.1°C
Dimensions (L x W x H) 185 x 72 x 36 mm
Weight (Metric) 300 g
Detection Range -2.00 to 16.00 pH
Temperature Compensation -20.0°C to +120.0°C
Battery Type 1.5V
Type Waterproof Portable pH Meter
Battery Life 200 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.