Learn More
Erlab Watertight Stainless Cable Grommet
Product Code. 31793970 Shop All Erlab Products
Change view
Click to view available options
Product Type:
Grommet
Unit Size:
Each
Product Code. Product Type Unit Size
31793970 Grommet Each
1 options
Product Code. 31793970 Supplier Erlab Supplier No. NORT000450
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket
The waterproof stainless steel cable gland facilitates cable routing whilst providing long-lasting protection against water ingress and environmental stress.
  • Service Fittings for Laboratory Enclosures: These service fittings allow water, gas, compressed air and electrical or data connections to be integrated directly inside Erlab enclosures, so that operators have all the utilities they need at the point of use, without compromising containment.
  • Safe Integration Inside the Working Volume:
    • Components designed for installation in Erlab enclosure panels
    • Sealed, leak-tight assembly that preserves the containment of the working volume
    • Corrosion-resistant materials suitable for laboratory environments
    • Positioning studied to keep the working surface clear and ergonomic
    • Electrical accessories compliant with applicable safety requirements
  • Practical, Ready-to-Install Solutions: Supplied as a complete kit with the required sealing and fixing components, these fittings can be installed on new equipment or retrofitted to an existing enclosure according to Erlab instructions.
  • Key Benefits:
    • Utilities available directly at the point of use.
    • Leak-tight installation that preserves enclosure containment.
    • Corrosion-resistant, easy-to-clean materials.
    • Ready-to-install kit designed for Erlab equipment.
    • Improved operator comfort and workstation efficiency.
Specifications
Product Type Grommet