missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ WAY 213613
GREENER_CHOICE

Product Code. 18043842
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18043842 10 mg 10mg
18045622 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18043842 Supplier Tocris Bioscience™ Supplier No. 2652/10
  • ONLINE EXCLUSIVE PRICE
    255.00 €
    240.45 € / 10mg
    Save 14.55 €
    5% Off
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent, non-substrate EAAT2 inhibitor

Potent, non-substrate inhibitor of EAAT2 (GLT-1) that displays > 44-fold selectivity over EAAT1 and EAAT3 (IC50 values are 85, 3787 and 5004 nM for EAAT2, EAAT3 and EAAT1 respectively). Exhibits no activity towards ionotropic and metabotropic glutamate receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868359-05-1
Molecular Formula C16H13BrF2N2O4

Specifications

Quantity 10 mg
Formula Weight 415.19
Purity >99%
Molecular Weight (g/mol) 415.191
Chemical Name or Material WAY 213613
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.