missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ WAY 316606 hydrochloride
GREENER_CHOICE

Product Code. 18782449
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18782449 10 mg 10mg
18780268 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18782449 Supplier Tocris Bioscience™ Supplier No. 4767/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Secreted Frizzled-related protein-1 (sFRP-1) inhibitor; increases Wnt signaling

WAY 316606 hydrochloride is a secreted Frizzled-Related Protein-1 (sFRP-1) inhibitor (IC50 = 0.65 μM); prevents sFRP-1 from interacting with Wnt and thus increases Wnt signaling. Increases total bone area in a murine calvarial organ culture assay.

We are aware of a study in PLOS Biology where WAY-316606 was found to stimulate hair follicle growth. Please note that WAY-316606 from Tocris is for laboratory research use only, and that we will only sell to established scientific businesses and institutes.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1781835-02-6
Quantity 10 mg
Product Type WAY 316606 hydrochloride
Purity >99%
Molecular Weight (g/mol) 484.94
Inhibitors WAY 316606 hydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.