Learn More
Tocris Bioscience™ WDR5 0103
GREENER_CHOICE
WDR5 antagonist
Brand:  Tocris Bioscience™ 5323/50
Encompass
Product Code. 15796706
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
WDR5 0103 is a WD repeat-containing protein 5 (WDR5) antagonist (Kd = 450 nM). Disrupts WDR5 interaction with MLL and inhibits MLL core complex methyltransferase activity in vitro. Has no effect on a panel of seven other methyltransferases, including SETD7.
Specifications
Specifications
>99%