Learn More
3M™ Welding Safety Goggles
Product Code. 15770028 Shop All 3M Products
Change view
Click to view available options
Lens Tint: Shade 5
Product Code.
15770028 Lens Tint: Shade 5
1 options
Product Code. 15770028 Supplier 3M™ Supplier No. 7100010431
  • ONLINE EXCLUSIVE PRICE
    342.00 €
    202.46 € / Pack of 10
    Save 139.54 €
    40% Off
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
3M™ Welding Safety Goggles feature a modern design to fit with respirator protection and have a sealed frame for additional protection against fine particles and gases.
  • Sealed design helps provide protection against liquids, dust, gases and vapors
  • Strong polycarbonate lens for impact resistance
  • Special anti-scratch and anti-fog coating for increased durability and improved vision
  • Wide nylon comfortable elastic strap for easy adjustability
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Approved, EN 166:2001
Lens Coating Anti-fog, Scratch-resistant
Description Sealed
Lens Tint Shade 5
Product Type Safety Goggles
Lens Material Polycarbonate