missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Wellwash™ Microplate Washer

Product Code. 15174725 Shop All Thermo Scientific Products
Change view
Click to view available options
Model:
Wellwash 1x12
Wellwash 1x8
Wellwash Versa 2x12
Wellwash Versa 2x8
Bottles:
Wash Bottle (2 L), Waste Bottle (2 L)
Wash Bottle (2 x 2 L), Rinse Bottle (1 x 2 L), Waste Bottle (1 x 4 L)
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Bottles Unit Size
15174725 Wellwash 1x12 Wash Bottle (2 L), Waste Bottle (2 L) Each
11708016 Wellwash 1x8 Wash Bottle (2 L), Waste Bottle (2 L) Each
11555485 Wellwash Versa 2x8 Wash Bottle (2 x 2 L), Rinse Bottle (1 x 2 L), Waste Bottle (1 x 4 L) Each
12000121 Wellwash Versa 2x8 Wash Bottle (2 x 2 L), Rinse Bottle (1 x 2 L), Waste Bottle (1 x 4 L) Each
15184725 Wellwash Versa 2x12 Wash Bottle (2 x 2 L), Rinse Bottle (1 x 2 L), Waste Bottle (1 x 4 L) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15174725 Supplier Thermo Scientific™ Supplier No. 5165040
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Wash 96- or 384-well plates securely with the Versa Microplate Washer, an easy-to-use microplate-strip washer for routine ELISA applications.

Wash 96- or 384-well plates securely with the Thermo Scientific™ Wellwash™ Versa Microplate Washer, an easy-to-use microplate-strip washer for routine ELISA applications. Designed for high performance and versatility, enjoy the ease of use and convenience provided through a graphical interface, local language version and USB port.

Simplify your ELISA workflow by using our eGuide (elisaguide.thermoscientific.com) to see our complete portfolio for all your ELISA needs.

Easy and convenient to use

  • Large color display and keypad for easy, convenient use
  • Clear display shows everything on one screen
  • User interface is logic and intuitive, allowing quick protocol development
  • Easy-to-use software requires minimal training for fast startup

Reliable and secure performance

  • Unpressurized bottles are safe and secure to use
  • Liquid level sensors are present in both wash and waste bottles
  • Automatic rinse and prime features assure safe performance
  • Aerosol cover prevents aerosolization of infectous diseases
  • Plate sensor recognizeds if plate is present

Optimal washing performance

  • Sweep mode guarantees an extremely low residual volume in the well, resulting in reliable assay results
  • Washing performance is further optimized by adjustable parameters such as dispense and aspiration height, and aspiration speed

Added versatility

  • A USB port for transferring protocols between several washers and for printing reports and log files
  • Specially designed 2x8 cell wash head is available for cell washing
  • Two 1x8 wash heads, two wash bottles and one rinse bottle, optional two 1x12 wash heads, 1x16 wash heads and 2x8 cell wash head offer increased flexibility
  • Possibility to connect to automation systems

Recommended for:

ELISA-based assays, cell washing

TRUSTED_SUSTAINABILITY

Specifications

Accuracy < 5% @ 300μL
Certifications/Compliance CE Marked
Connections USB
Wash/Waste Bottle Capacity 1
Bottles Wash Bottle (2 L), Waste Bottle (2 L)
Precision < 3% (CV) @ 300μL (96-well plate)
Dispense Volume 50 to 400 μL
Wash Heads 1 x 12
Plate Type 96-well Plate
Residual Volume <2 μL
Weight (Metric) 8 kg
Wash Volume 50 to 1000 μL
Display LCD
For Use With (Application) ELISA-Based Assays
Frequency 50/60 Hz
Model Wellwash 1x12
Cell Washing No
Includes 1 x 12 wash head, 1 x 2L wash bottle, 1 x 2L waste bottle, aerosol cover
Width (English) 15.2 in.
Width (Metric) 385 mm
Height (English) 13.6 in.
Height (Metric) 345 cm
Depth (English) 9.4 in.
Depth (Metric) 240 mm
Automation Interface No
Weight (English) 17.6 lb.
Electrical Requirements 100/240 V 50/60 Hz
Voltage 100/240 V
Prime Volume 5 to 100 mL
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.