missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam Western Blot Marker Pen, For PVDF And Nitrocellulose, 1 Ea.
Description
The Western Blot Marker Pen is designed for annotating western blots and marking the positions of molecular weight standard markers on PVDF or nitrocellulose membranes.
- Constituents: 10% Glycerol and 1% C.I. Acid Blue 83
- Storage: Store at +4°C
- Shipping: Blue Ice
Specifications
Specifications
| Format | Pen |
| Form | Liquid |
| Shipping Condition | Blue Ice |
| Membrane Compatibility | Nitrocellulose, PVDF |
| Quantity | 1 Ea. |
| For Use With (Application) | Western Blot |
| Storage Requirements | Store at 4°C. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?