missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam Western Blot Stripping Buffer, Liquid, 500 mL
Description
- Form: Liquid
- Storage: Store at Ambient
- Shipping: Ambient - Cannot Ship with Ice
- pH: 2.9 to 3.1
- Constituents: 88% Water, 10% Sodium lauryl sulfate
- Application: Uniformly removes antibodies from Western blots for reprobing
Specifications
Specifications
| Product Type | Wash Solution |
| Buffer | 10% sodium lauryl sulfate, pH 2.9 to 3.1 |
| Content And Storage | Store at ambient temperature. |
| Format | Bottle |
| Form | Liquid |
| Shipping Condition | Ambient |
| Membrane Compatibility | Nitrocellulose, PVDF |
| Formulation | pH 2.9 to 3.1, 88% Water, 10% Sodium lauryl sulfate |
| Quantity | 500 mL |
| For Use With (Application) | Western Blot |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?