missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WesternDot™ 625 Western Blot Kits

Product Code. 10062623
Change view
Click to view available options
Reactivity:
Mouse
Rabbit
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Reactivity unitSize
10062623 Mouse Each
10469433 Rabbit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10062623 Supplier Invitrogen™ Supplier No. W10132

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The WesternDot 625 Western Blot kits combine the unique properties of Qdot 625 nanocrystals with the high-affinity streptavidin-biotin interaction to allow simple, highly sensitive detection of proteins on western blots.

The WesternDot 625 Western Blot kits combine the unique properties of Qdot 625 nanocrystals with the high-affinity streptavidin-biotin interaction to allow simple, highly sensitive detection of proteins on western blots. The WesternDot kits contain optimized, ready-to-use or ready-to-dilute reagents for sensitive immunodetection of proteins immobilized on nitrocellulose (NC) or polyvinylidene difluoride (PVDF) membranes.

WesternDot 625 Western Blot kits utilize extremely bright Qdot 625 nanocrystals and include reagents for sensitive imaging of proteins probed with mouse or rabbit primary antibodies.

Features include:
• Detection performance and ease-of-use exceed existing ECL-based methods
• Make use of extremely bright Qdot 625 streptavidin conjugate to allow direct imaging of blots while avoiding the need to spend time and money on optimizing multiple exposures to film in the dark
• Contains reagents sufficient for 20 mini-gel blots (7 x 10 cm/blot)

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • WesternDot™ Blocking Buffer, Component A
• Wash Buffer, Component B
• Biotin-XX Goat Anti-Mouse IgG (H+L), Component C
• Qdot™ 625 Streptavidin Conjugate, Component D
• WesternDot™ Staining Dish, Component E

Store at 4°C, Do Not Freeze.
Detection Location In-Blot Detection
Color Red
Detection Method Fluorescence
Label or Dye Qdot 625
Label Type Qdot Nanocrystals
Quantity 1 kit
Reactivity Mouse
Sample Type Protein, Cell Extracts
Shipping Condition Room Temperature
Product Type 625 Goat Anti-Mouse Western Blot Kit
For Use With (Equipment) iBlot, Photographic Imager
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.