missing translation for 'onlineSavingsMsg'
Learn More

Cytiva WFI Quality Water

Product Code. 15939355
Change view
Click to view available options
:
Water for Injection; Packaging: Single-Use Bag
Product Code.
15939355 Water for Injection; Packaging: Single-Use Bag
1 options
This item is not returnable. View return policy

Product Code. 15939355

Brand: Cytiva CH30154.01

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Water for injection (WFI) quality water is widely used in the pharmaceutical industry. Today’s rapid advances in biotechnology and biopharmaceutical manufacturing create an ever-increasing need for dependable sources of WFI quality water.

  • HyClone HyPure WFI Quality Water:
    • Meets stringent Pharmacopeial specifications
    • Manufactured in an ISO 9001 certified facility
    • Certificate of analysis (CoA) for each lot of water produced
    • Low levels of endotoxin
    • Sterile filtered
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Conductivity < 1.1 μS/cm at 20°C to 25°C
Endotoxin < 0.25 EU/mL
Packaging Single-Use Bag
Residue after Evaporation Not More Than 3 mg (0.003%)
Sterility Sterile-filtered
Quantity 10 L
Purification Method Reverse Osmosis/Distillation
For Use With (Application) Hydration of Cell Culture Media, Supplements, and Salt Solutions; Rinsing of Primary Packaging, Production Vessels, Equipment, and Room Surfaces; Reconstitution and Rehydration of Product During Synthesis, Preparation of Buffers for Chromatographic Purification
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.