missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Anodisc™ Filter Membranes

Product Code. 11334734
Change view
Click to view available options
Pore Size:
0.02 μm
0.1 μm
0.2 μm
Diameter (Metric):
13 mm
25 mm
47 mm
Includes:
No Support Ring
Support Ring
Unit Size:
Pack of 100
Pack of 50
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Diameter (Metric) Includes unitSize
11334734 0.2 μm 13 mm No Support Ring Pack of 100
11324734 0.1 μm 13 mm No Support Ring Pack of 100
11314734 0.02 μm 13 mm No Support Ring Pack of 100
11335044 0.1 μm 25 mm Support Ring Pack of 50
10480713 0.02 μm 25 mm Support Ring Pack of 50
11304734 0.2 μm 25 mm Support Ring Pack of 50
11384724 0.2 μm 47 mm Support Ring Pack of 50
11374724 0.1 μm 47 mm Support Ring Pack of 50
11364724 0.02 μm 47 mm Support Ring Pack of 50
11954416 0.2 μm 47 mm No Support Ring Pack of 50
11345044 0.02 μm 47 mm No Support Ring Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 11334734 Supplier Cytiva Supplier No. 68097023
Add to basket
Add to basket
This item is not returnable. View return policy

Constructed with precision pore structure and narrow pore size distribution. Whatman™ Anodisc™ Filter Membranes ensure a high level of particle removal efficiency. Captures microorganisms and particulates on the surface of the membrane for subsequent analysis.

Anopore inorganic membrane circles available with or without support ring contain no lateral crossovers between individual pores.
  • Wide solvent compatibility reduces the need to stock a variety of membranes in the laboratory.
  • No additives used in the manufacturing process ensures minimal extractables and no sample contamination.
  • Extremely low protein binding minimizes sample loss.
  • Virtually transparent when wet making it highly suitable for microscopy studies.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Material Aluminum Oxide
Pore Size 0.2 μm
Hydrophobicity Hydrophilic
Includes No Support Ring
Membrane Type Anodisc 13
Quantity 100 Pack
Diameter (Metric) 13 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.