missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Anotop™ 25 Sterile Syringe Filters

Remove virtually 100% of particulates larger than pore size

Brand:  Cytiva 6809-2102

840.60 EUR valid until 2024-05-06
Use promo code "21711" to get your promotional price.



Code : 75

Additional Details : Weight : 0.38700kg

 View more versions of this product

Product Code. 11353155

  • 920.00 € / Pack of 50
Estimated Shipment: 02-07-2024
to see stock.
Add to basket

Description

Description

Remove virtually 100% of particulates larger than pore size

Compatible with aqueous suspensions and a wide range of organic solvents. Handle volumes 100mL (25mm diameter filter).

  • Nondeformable, honeycombed Anopore™ membrane
  • Inherently hydrophilic membrane is free of adhesives and surfactants
  • Low protein binding, low fluid retention for maximum sample recovery
  • Unidirectional flow

Order Info

Hexagonal, thermal-welded polypropylene housing is free of adhesives and binders. Sterile units are individually packaged to avoid contamination.

Specifications

Specifications

Anotop 25 mm Sterile Syringe Filters, 0.02 μm (50 pcs)
Anapore
Sterile
HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water
Polypropylene (PP)
Female luer lock
6.9 bar (100 psi)
113°F
Polypropylene
100 mL (25 mm Diameter Filter)
Single-use
0.02 μm
4.78 cm2
<150 μL
Hydrophilic
Male luer
50/Pk.
45°C
Syringe Filter
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers