missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Anotop™ 25 Sterile Syringe Filters
Remove virtually 100% of particulates larger than pore size
920.00 € - 970.00 €
Specifications
Disposable | Single-use |
---|---|
Material | Anapore |
Sterility | Sterile |
Filtration Area | 4.78 cm2 |
For Use With (Application) | HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
11384714
|
Cytiva
6809-2122 |
0.2 μm |
955.00 €
Pack of 50 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
11353155
|
Cytiva
6809-2102 |
0.02 μm |
920.00 €
Pack of 50 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
11374714
|
Cytiva
6809-2112 |
0.1 μm |
970.00 €
Pack of 50 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
Remove virtually 100% of particulates larger than pore size
Compatible with aqueous suspensions and a wide range of organic solvents. Handle volumes 100mL (25mm diameter filter).
- Nondeformable, honeycombed Anopore™ membrane
- Inherently hydrophilic membrane is free of adhesives and surfactants
- Low protein binding, low fluid retention for maximum sample recovery
- Unidirectional flow
Order Info
Hexagonal, thermal-welded polypropylene housing is free of adhesives and binders. Sterile units are individually packaged to avoid contamination.
Specifications
Single-use | |
Sterile | |
HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water | |
Polypropylene (PP) | |
Male luer | |
50/Pk. | |
45°C | |
Syringe Filter |
Anapore | |
4.78 cm2 | |
<150 μL | |
Female luer lock | |
6.9 bar (100 psi) | |
113°F | |
Polypropylene | |
25 mm |