Learn More
Cytiva Whatman™ Anotop™ 25 Sterile Syringe Filters
Remove virtually 100% of particulates larger than pore size
Brand: Cytiva 6809-2112
Code : 75
Additional Details : Weight : 0.40000kg
Description
Remove virtually 100% of particulates larger than pore size
Compatible with aqueous suspensions and a wide range of organic solvents. Handle volumes 100mL (25mm diameter filter).
- Nondeformable, honeycombed Anopore™ membrane
- Inherently hydrophilic membrane is free of adhesives and surfactants
- Low protein binding, low fluid retention for maximum sample recovery
- Unidirectional flow
Order Info
Hexagonal, thermal-welded polypropylene housing is free of adhesives and binders. Sterile units are individually packaged to avoid contamination.
Specifications
Anotop 25 mm Sterile Syringe Filters, 0.1 μm (50 pcs) | |
Anapore | |
Sterile | |
HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water | |
Polypropylene (PP) | |
Male luer | |
50/Pk. | |
45°C | |
Syringe Filter |
Single-use | |
0.1 μm | |
4.78 cm2 | |
<150 μL | |
Female luer lock | |
6.9 bar (100 psi) | |
113°F | |
Polypropylene | |
25 mm |