Learn More
Cytiva Whatman™ Antibiotic Assay Discs
Product Code. 11340654
Change view
Click to view available options
Diameter (Metric):
6 mm
13 mm
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11340654 6 mm Pack of 1000
11350654 13 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11340654 Supplier Cytiva Supplier No. 2017006
  • ONLINE EXCLUSIVE PRICE
    259.00 €
    186.00 € / Pack of 1000
    Save 73.00 €
    28% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Used in conjunction with the Kirby-Bauer method of antibiotic testing. Whatman™ Antibiotic Assay Discs determine the type of causal agent of infectious diseases and their sensitivity to antibiotics and chemotherapeutic agents.
Antibiotic assay discs are used for determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method.
  • For use with Kirby-Bauer method of antibiotic testing
  • Allows rational and selective chemotherapy
TRUSTED_SUSTAINABILITY
Specifications
Product Type Antibiotic Assay Discs
Test Type Infectious Disease, Antibiotic Susceptibility Sensitivity
Packaging Type 1000 Discs/Pk.
Quantity 1000 Discs/Pk.
Diameter (Metric) 6 mm