Learn More
Cytiva Whatman™ Carbon-Cap Disposable Activated Carbon Capsules
Filled with high-purity, high-efficiency, acid-washed, granular-activated carbon. Whatman™ Carbon Cap Disposable Activated Carbon Capsules are suitable for adsorption of organics from air and removal of color, organics and chlorine from water.
Brand: Cytiva 6704-1500
Code : 75
Additional Details : Weight : 0.31772kg
Description
Filled with high-purity, high-efficiency, acid-washed, granular-activated carbon. Whatman™ Carbon Cap Disposable Activated Carbon Capsules are suitable for adsorption of organics from air and removal of color, organics and chlorine from water.
- Remove organic odors, color, and colloids from water and air
- Activated carbon plus a pleated HEPA filter media retains 99.97% of all particles 0.3μm
- Maximum housing pressure: 4.1 bar (60psi.)
- Carbon-Cap 75 offers 26,000m2 activated carbon surface area
- Carbon-Cap 150 offers 82,000m2 filter area equivalent
Applications:
For vacuum pump outlet removal of smoke and oil.
Specifications
Carbon Cap 150, capsule filter with activated carbon (1 pc) | |
For water, chemical, and reagent purification | |
4.1 bar (60 psi) |
82,000 m2 | |
Polypropylene (PP) |