Learn More
Cytiva Whatman™ Drying Rack for 903 Protein Saver Cards
Product Code. 11909134
Change view
Click to view available options
Includes:
Hook-and-Loop Mounting Sticker
Mounting Sticker not included
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
11909134 Hook-and-Loop Mounting Sticker Pack of 10
11359394 Mounting Sticker not included Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11909134 Supplier Cytiva Supplier No. 10539521
  • ONLINE EXCLUSIVE PRICE
    92.40 €
    86.59 € / Pack of 10
    Save 5.81 €
    6% Off
View all promotions
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Air-dries blood specimens in a suspended horizontal position. Whatman™ Drying Rack for 903 Protein Saver Card is easily assembled and can be affixed to a wall or counter top with optional hook-and-loop stickers.
Air-dries blood specimens in a suspended horizontal position. Whatman™ Drying Rack for 903 Protein Saver Card is easily assembled and can be affixed to a wall or counter top with optional hook-and-loop stickers.
  • For use with neonatal human diagnostic testing cards
  • CE Marked
  • Accommodate multiple collection forms at one time
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 903 Protein Saver Card
Quantity 10 Pack
Includes Hook-and-Loop Mounting Sticker
Product Type Drying Rack