Learn More
QIAGEN EasiCollect
For simple, reliable and standardized sample collection.
Brand:  QIAGEN WB120462
Code : NEW
Additional Details : Weight : 1.09416kg
Product Code. 15550143
  • 275.00 € / Pack of 50
Estimated Shipment: 17-06-2024
to see stock.
Description
Description
For simple, reliable and standardized sample collection.
50 buccal sample collection devices for simplifies sample uptake and transportation.
Specifications
Specifications
Buccal Cells, Saliva
Includes an Indicating FTA Card that changes color from pink to white upon sample addition
DNA Sample Collection Device
50 Pk.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning