missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ High-Purity Glass Thimbles

Product Code. 11390384
Change view
Click to view available options
Length (Metric):
90 mm
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11390384 90 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11390384 Supplier Cytiva Supplier No. 2814259
  • ONLINE EXCLUSIVE PRICE
    222.00 €
    205.00 € / Pack of 25
    Save 17.00 €
    7% Off
 Restricted Item
Estimated Shipment: 02-04-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use for pollution monitoring when analysis of hot and acidic gases is required. Whatman™ High-Purity Thimbles are designed to fit most Soxhlet extraction units and can be used with solvents that are incompatible with cellulose thimbles.

Features 

  • Free of binders and additives
  • Tapered
  • Dimensions: 25 × 90mm
  • Nominal particle retention rating: 0.8μm 

 High-Purity Glass

  • 100% borosilicate glass microfiber (HP-GF)
  • Resistant to temperatures up to 500°C
 Applications
  • Analysis of hot and acidic gases during emission control and smoke stack gas monitoring
  • Analysis of pesticide residues using Soxhlet extraction
  • Determination of oil/fat content of foods
  • Analysis of oil and grease in solid wastes
TRUSTED_SUSTAINABILITY

Specifications

Description GE’s Whatman 100% borosilicate glass microfiber (HP-GF) thimbles are designed to fit most Soxhlet extraction units. They are well-suited to pollution monitoring when analysis of hot and acidic gases is required.
Diameter (English) Inside 0.98 in.
Diameter (Metric) Inside 25 mm
Diameter (Metric) Outer 25 mm
Material Borosilicate Glass Fibers
Wall Thickness 1.7 mm
For Use With (Application) Smoke stack gas monitoring. Soxhlet extraction. Analyzing pesticide residues. Determining oil/fat content of foods. Analysis of oil and grease in solid wastes.
Grade HP-GF
For Use With (Equipment) For soxhlet extractor
Length (English) 3.54 in.
Length (Metric) 90 mm
Max. Temperature (Metric) 500°C
Product Type Extraction Thimble
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • FILTER GLASS CIRCLE
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: glass, oxide, chemicals
  • UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.