missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QIAGEN QIAcard FTA Elute Indicating Micro
For collection, stabilization, processing, transport and archiving of biological samples. 1 sample area per card.
160.00 € - 885.00 €
Specifications
Content And Storage | Room Temperature |
---|---|
Description | Collect, process, transport, and store colorless biological samples, such as buccal or saliva samples. With FTA Elute, proteins remain tightly bound while DNA is eluted from the matrix. |
For Use With (Application) | Multiplex PCR, Sequencing after PCR amplification, SNP analysis, STR analysis, HLA typing, Whole genome amplification, Quantitative PCR, Biobanking, Pharmacogenomics, Genotyping, Genetic identification, Transgenic detection |
Includes | Inert dye |
Product Type | Indicating FTA Elute Micro Card |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12445432
|
QIAGEN
WB120412 |
25 Pk. |
160.00 €
Pack of 25 |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11904426
|
QIAGEN
WB120411 |
100 Pk. |
885.00 €
Each |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- FTA Elute technology for convenient water based elution of nucleic acids.
- QIAcard Indicating FTA Cards include a pink dye that turns white when a colorless sample, e.g. buccal cells or saliva is applied.
- Stabilization and protection of nucleic acids at room temperature with for spot volumes of 125 μL.
- Cell lysis on contact, denatures proteins and protects DNA from degradation.
Specifications
Room Temperature | |
Multiplex PCR, Sequencing after PCR amplification, SNP analysis, STR analysis, HLA typing, Whole genome amplification, Quantitative PCR, Biobanking, Pharmacogenomics, Genotyping, Genetic identification, Transgenic detection | |
Indicating FTA Elute Micro Card | |
15 to 30 min |
Collect, process, transport, and store colorless biological samples, such as buccal or saliva samples. With FTA Elute, proteins remain tightly bound while DNA is eluted from the matrix. | |
Inert dye | |
40 μL |