missing translation for 'onlineSavingsMsg'
Learn More

Cytiva™ Whatman™ Lens Cleaning Tissue

Product Code. 11360654
Change view
Click to view available options
Dimensions (L x W):
150 x 100 mm
300 x 200 mm
570 x 460 mm
Quantity:
100 Sheets
25 Sheets/Wallet, 25 Wallets/Cs.
Unit Size:
Pack of 100
Pack of 500
Pack of 625
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Quantity Unit Size
11360654 150 x 100 mm 25 Sheets/Wallet, 25 Wallets/Cs. Pack of 625
10149470 300 x 200 mm 100 Sheets Pack of 100
11380654 570 x 460 mm 100 Sheets Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11360654 Supplier Cytiva™ Supplier No. 2105841
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Provides scratch-free cleaning solutions. Whatman™ Lens Cleaning Tissue safely removes surface moisture and grease.

Soft textured strong tissue leaves no fibers. Protects lenses and other optical surfaces made from glass, quartz, or plastic from scratches.
  • Chemically pure, free of silicone and other additives
  • High absorbency leads to increased safety upon removal of surface moisture and grease
  • Thickness 0.035 to 0.040mm
  • Grade 105 sheets
TRUSTED_SUSTAINABILITY

Specifications

Product Type Lens Wipe
Description Won't damage lenses or optical surfaces, leaves no fibers
Dimensions (L x W) 150 x 100 mm
For Use With (Application) Safe removal of surface moisture and grease.
Quantity 25 Sheets/Wallet, 25 Wallets/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.