Learn More
Cytiva Whatman™ Litmus Blue Test Paper
Impregnated with a mixture of pH indicators, acid-alkali litmus papers assume a color specific to sample pH upon contact with a sample. Whatman™ Litmus Blue Test Paper provides rapid determination of pH values and has a variety of applications in laboratories and industry.
Brand:  Cytiva 2600-201A
Additional Details : Weight : 0.01800kg
 View more versions of this product
Product Code. 11380674
  • 19.75 € / Each
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
Description
Description
Impregnated with a mixture of pH indicators, acid-alkali litmus papers assume a color specific to sample pH upon contact with a sample. Whatman™ Litmus Blue Test Paper provides rapid determination of pH values and has a variety of applications in laboratories and industry.
Mixed-indicator tests designed for fast, accurate, and reliable pH solution testing.
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Convenient and portable for field use
  • Color change in pH solutions ranging from approximately 5 to 8
  • Economical
Specifications
Specifications
Blue
Reel
Test Paper
5 to 8
1 Each