missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Litmus Blue Test Paper

Impregnated with a mixture of pH indicators, acid-alkali litmus papers assume a color specific to sample pH upon contact with a sample. Whatman™ Litmus Blue Test Paper provides rapid determination of pH values and has a variety of applications in laboratories and industry.

19.75 € - 21.57 €

Specifications

Color Indicator Blue
pH Range 5 to 8
Type Test Paper
View More Specs

Products 2
Product Code Brand Format Price Quantity & Availability  
Product Code Brand Format Price Quantity & Availability  
11380674
View Documents Promotion Details
Cytiva
2600-201A
Reel
19.75 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
 
11334974
View Documents Promotion Details
Cytiva
10360300
Strip
21.57 €
Pack of 100
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
Description

Description

Impregnated with a mixture of pH indicators, acid-alkali litmus papers assume a color specific to sample pH upon contact with a sample. Whatman™ Litmus Blue Test Paper provides rapid determination of pH values and has a variety of applications in laboratories and industry.

Mixed-indicator tests designed for fast, accurate, and reliable pH solution testing.
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Convenient and portable for field use
  • Color change in pH solutions ranging from approximately 5 to 8
  • Economical
Specifications

Specifications

Blue
5 to 8
Test Paper
Videos
SDS
Documents

Documents

Product Certifications