missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ 0.45 μm Pore Size (ME 25) Mixed Cellulose Ester Membranes, White/Black Grid

Product Code. 11355364
Change view
Click to view available options
Diameter (Metric):
50 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11355364 50 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11355364 Supplier Cytiva Supplier No. 10406572
  • ONLINE EXCLUSIVE PRICE
    157.00 €
    151.00 € / Pack of 100
    Save 6.00 €
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Whatman™ (ME 25) Mixed Cellulose Ester Membranes, White/Black Grid are particularly effective in applications requiring higher flow rates and larger volume filtration. Supplied with clearly defined grid lines at regularly spaced intervals to assist in microbiological colony counting.

  • Material: Mixed Cellulose Ester
  • Material (Membrane): ME 25
  • Pore Size: 0.45 μm
  • Size (Metric) Grid: 5 mm
  • Color: White
  • Grid Color: Black
  • Autoclavable: Yes
  • Sterility: Sterile
  • Diameter (Metric): 50 mm
  • Flow Rate: 12.5 sec./100 mL/12.5 cm² (at Δp = 0.9 bar)
  • For Use With (Application): Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Color White
Filter Type Gridded Filter
Material MCE
Material (Membrane) ME 25
Pore Size 0.45 μm
Sterility Sterile
Flow Rate 12.5 sec./100 mL/12.5 cm² (at Δp = 0.9 bar)
For Use With (Application) Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Grid Color Black
Diameter (English) 1.97 in.
Diameter (Metric) 50 mm
Show More Show Less
compliance-icons
Product Identifier
  • ME25/20ST, 0.45µm 50mm
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 1
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: nitrocellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.