Learn More
Cytiva Whatman™ Mini-UniPrep™ Syringeless Filters
Processes sample loads in one-third the time. Whatman™ Mini-UniPrep™ Syringeless Filters provide a faster and easier way to remove particulates from samples being prepared for HPLC and UHPLC analysis.
Brand: Cytiva UN503NPUGMF
2318.00 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Code : 50
Additional Details : Weight : 0.22000kg
Description
Processes sample loads in one-third the time. Whatman™ Mini-UniPrep™ Syringeless Filters provide a faster and easier way to remove particulates from samples being prepared for HPLC and UHPLC analysis.
Whatman Mini-UniPrep™ Transparent Syringeless Filters quickly and easily remove particulates from samples being prepared for HPLC/UHPLC analysis, while maintaining sample visibility.
- Pre-assembled all-in-one filtration unit eliminates the need for syringe filter, syringe, vial and septum, reducing costs by up to 40%
- Wide range of membrane choices in 0.2 and 0.45 μm pore sizes to meet specific sample application requirements
- Compatible with most major autosamplers for high-throughput analysis
- Fewer consumables required
- Plunger contains a filtration membrane at one end and a pre-attached cap/septum at the other
- Can be used either manually or with a compressor unit
- Multi-sample compressor processes up to 6 samples at a time, further improving sample processing time and reducing hand stress
- Designed to fit into any autosampler accommodating 12 x 32mm vials -OR-
Septum can be pierced with needle and sample drawn off for manual injection into an analyzer
Specifications
Translucent | |
Single-use | |
PTFE-coated Silicone Rubber Septa | |
0.45 μm | |
Routine HPLC/UHPLC analysis; Sample filtration; Composite assays; Content uniformity; Protein precipitation; Solubility/dissolubility testing; Ion chromatography | |
Polypropylene (PP) | |
1000/Pk. | |
50°C | |
0.4 mL |
Equivalent in size to 12 x 32mm vials | |
Autosampler, Compressor | |
Glass Microfiber | |
0.28 cm2 | |
50 μL | |
Vial with incorporated filtration membrane, plunger and cap/septa | |
0.6 bar / 8 psi (to compress) | |
Syringeless Filter |