Learn More
Cytiva Whatman™ Mini-UniPrep™ Syringeless Filters
Processes sample loads in one-third the time. Whatman™ Mini-UniPrep™ Syringeless Filters provide a faster and easier way to remove particulates from samples being prepared for HPLC and UHPLC analysis.
269.00 € - 2880.00 €
Specifications
For Use With (Application) | Routine HPLC/UHPLC analysis; Sample filtration; Composite assays; Content uniformity; Protein precipitation; Solubility/dissolubility testing; Ion chromatography |
---|---|
Type | Syringeless Filter |
Product Code | Brand | Material (Membrane) | Pore Size | No. per Pack | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material (Membrane) | Pore Size | No. per Pack | Price | Quantity & Availability | |||
11338744
|
Cytiva
UN203NPUAQU |
PVDF | 0.45 μm | 100/Pk. |
269.00 €
Pack of 100 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||
11348744
|
Cytiva
UN203NPUDPP |
Polypropylene Depth Filter | 0.45 μm | 100/Pk. |
269.00 €
Pack of 100 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||
11388744
|
Cytiva
UN203NPUPES |
PES | 0.45 μm | 100/Pk. |
278.00 €
Pack of 100 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||
11398754
|
Cytiva
UN503NPUNYL |
Nylon | 0.45 μm | 1000/Pk. |
2440.00 €
Pack of 1000 |
Estimated Shipment: 25-07-2024
Log in to see stock. |
|||
11378744
|
Cytiva
UN203NPUORG |
PTFE | 0.45 μm | 100/Pk. |
269.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
11318764
|
Cytiva
UN503NPUPES |
PES | 0.45 μm | 1000/Pk. |
2525.00 €
Pack of 1000 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11368744
|
Cytiva
UN203NPUNYL |
Nylon | 0.45 μm | 100/Pk. |
269.00 €
Pack of 100 |
Estimated Shipment: 25-07-2024
Log in to see stock. |
|||
11328754
|
Cytiva
UN503NPENYL |
Nylon | 0.2 μm | 1000/Pk. |
2440.00 €
Pack of 1000 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
11378754
|
Cytiva
UN503NPUDPP |
Polypropylene Depth Filter | 0.45 μm | 1000/Pk. |
2525.00 €
Pack of 1000 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11398734
|
Cytiva
UN203NPENYL |
Nylon | 0.2 μm | 100/Pk. |
333.00 €
Pack of 100 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||
11368754
|
Cytiva
UN503NPUAQU |
PVDF | 0.45 μm | 1000/Pk. |
2440.00 €
Pack of 1000 |
Estimated Shipment: 25-07-2024
Log in to see stock. |
|||
11338754
|
Cytiva
UN503NPEORG |
PTFE | 0.2 μm | 1000/Pk. |
2440.00 €
Pack of 1000 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||
11308764
|
Cytiva
UN503NPUORG |
PTFE | 0.45 μm | 1000/Pk. |
2440.00 €
Pack of 1000 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||
11358744
|
Cytiva
UN203NPUGMF |
Glass Microfiber | 0.45 μm | 100/Pk. |
269.00 €
Pack of 100 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
11308744
|
Cytiva
UN203NPEORG |
PTFE | 0.2 μm | 100/Pk. |
292.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
11388754
|
Cytiva
UN503NPUGMF |
Glass Microfiber | 0.45 μm | 1000/Pk. |
2440.00 €
Pack of 1000 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
11318754
|
Cytiva
UN503NPEAQU |
PVDF | 0.2 μm | 1000/Pk. |
2440.00 €
Pack of 1000 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
11388734
|
Cytiva
UN203NPEAQU |
PVDF | 0.2 μm | 100/Pk. |
269.00 €
Pack of 100 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||
11318744
|
Cytiva
UN203NPEPES |
PES | 0.2 μm | 100/Pk. |
301.00 €
Pack of 100 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||
11328764
|
Cytiva
UN503NPUPP |
Polypropylene | 0.45 μm | 1000/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11398744
|
Cytiva
UN203NPUPP |
Polypropylene | 0.45 μm | 100/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Processes sample loads in one-third the time. Whatman™ Mini-UniPrep™ Syringeless Filters provide a faster and easier way to remove particulates from samples being prepared for HPLC and UHPLC analysis.
Whatman Mini-UniPrep™ Transparent Syringeless Filters quickly and easily remove particulates from samples being prepared for HPLC/UHPLC analysis, while maintaining sample visibility.
- Pre-assembled all-in-one filtration unit eliminates the need for syringe filter, syringe, vial and septum, reducing costs by up to 40%
- Wide range of membrane choices in 0.2 and 0.45 μm pore sizes to meet specific sample application requirements
- Compatible with most major autosamplers for high-throughput analysis
- Fewer consumables required
- Plunger contains a filtration membrane at one end and a pre-attached cap/septum at the other
- Can be used either manually or with a compressor unit
- Multi-sample compressor processes up to 6 samples at a time, further improving sample processing time and reducing hand stress
- Designed to fit into any autosampler accommodating 12 x 32mm vials -OR-
Septum can be pierced with needle and sample drawn off for manual injection into an analyzer
Specifications
Routine HPLC/UHPLC analysis; Sample filtration; Composite assays; Content uniformity; Protein precipitation; Solubility/dissolubility testing; Ion chromatography | |
Syringeless Filter |