missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Mixed Cellulose Ester Membranes - WME Range, 0.45 μm Pore Size

Product Code. 11914544
Change view
Click to view available options
Surface Type:
Grid
Plain
Sterility:
Non-sterile
Sterile
Includes:
Interleaving paper
Pad
Pad, Interleaving paper
Quantity:
100 Pack
1000 Pack
200 Pack
Unit Size:
Pack of 100
Pack of 1000
Pack of 200
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Surface Type Sterility Includes Quantity unitSize
11914544 Grid Sterile Interleaving paper 1000 Pack Pack of 1000
10153971 Grid Non-sterile Pad 100 Pack Pack of 100
11374824 Plain Sterile Pad 100 Pack Pack of 100
11929207 Grid Sterile Interleaving paper 200 Pack Pack of 200
10030461 Grid Sterile Pad, Interleaving paper 100 Pack Pack of 100
10756262 Grid Sterile Interleaving paper 100 Pack Pack of 100
11324834 Grid Non-sterile Interleaving paper 100 Pack Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 11914544 Supplier Cytiva Supplier No. 7141154
  • ONLINE EXCLUSIVE PRICE
    992.00 €
    955.00 € / Pack of 1000
    Save 37.00 €
    3% Off
View all promotions
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Composed of cellulose acetate and cellulose nitrate membranes. Whatman™ Mixed Cellulose Ester Membranes - WME Range are characterized by a smoother and more uniform surface than pure nitrocellulose filters.

  • Gridded surface (3.1mm intervals) make filters ideal for colony counting after incubation
  • Color contrast provided by the filter surface also facilitates particle detection and minimizes eye fatigue.
  • Special nontoxic ink used is completely free from bacterial growth inhibitors
  • Manufactured under strictly controlled cleanroom conditions

Order Info

Autoclavable pack has 10 envelopes of 10 filters/pads for sterilization in the microbiological laboratory.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Color White
Material (Membrane) MCE
Pore Size 0.45 μm
Sterility Sterile
For Use With (Application) Microbiological work, sample preparation, microbiological studies, filtration of aqueous solutions
Grid Color Black
Includes Interleaving paper
Packaging Individually Packed
Protein Binding Capacity Medium
Quantity 1000 Pack
Size (Metric) Grid 3.1mm
Surface Type Grid
Temperature (English) Operating 266°F
Temperature (Metric) Operating 130°C
Diameter (English) 1.85 in.
Diameter (Metric) 47 mm
Show More Show Less
compliance-icons
Product Identifier
  • WME, white/black grid 3.1mm, sterile, without pad, 0.45µm, 47mm
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 1
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: nitrocellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.