missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ pH Indicators

Product Code. p-4372195
Click to view available options
pH Range:
0 to 14
1 to 11
2 to 9
Quantity:
1 Roll
100 Pack
200 Pack
Unit Size:
Each
Pack of 100
Pack of 200
This item is not returnable. View return policy

Product Code. 11354974

Brand: Cytiva 10362000

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Provides rapid determination of pH values. Whatman™ pH Indicators are impregnated with a mixture of several indicators that change color when they come in contact with a particular sample solution.

Determine pH using the supplied color comparison table. Test strips assume a particular color on contact with the sample solution.

  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Inexpensive
  • Convenient and portable for field use. These pH indicators have been impregnated with a mixture of several indicators
TRUSTED_SUSTAINABILITY

Specifications

pH Range 0 to 14
Format Strip
Includes Color comparison chart
Quantity 100 Pack
Type Indicator Paper
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.