missing translation for 'onlineSavingsMsg'
Learn More

Cytiva™ Whatman™ pH Indicators Reel Dispensers

Product Code. 11340674
Change view
Click to view available options
pH Range:
0.5 to 5.5
1 to 11
1 to 14
4 to 7
6.4 to 8
8 to 10
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
11340674 0.5 to 5.5 Each
11360674 6.4 to 8 Each
11330674 1 to 14 Each
10052251 1 to 11 Each
11350674 4 to 7 Each
11370674 8 to 10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11340674 Supplier Cytiva™ Supplier No. 2600101A
  • ONLINE EXCLUSIVE PRICE
    21.45 €
    19.70 € / Pack of 1
    Save 1.75 €
    8% Off
View all promotions
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for rapid determination of pH values

  • Instant pH readings
  • Accurate for wide range of routine pH testing
  • Convenient and portable for field use

TRUSTED_SUSTAINABILITY

Specifications

pH Range 0.5 to 5.5
Format Reel
For Use With (Application) Routine pH testing
Type pH Indicator Dispenser
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.