missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ pH Indicators Reel Dispensers
Used for rapid determination of pH values
21.20 € - 33.75 €
Specifications
For Use With (Application) | Routine pH testing |
---|---|
Type | pH Indicator Dispenser |
Product Code | Brand | pH Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH Range | Price | Quantity & Availability | |||||
11340674
|
Cytiva
2600-101A |
0.5 to 5.5 |
21.20 €
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
11360674
|
Cytiva
2600-103A |
6.4 to 8 |
21.20 €
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
11330674
|
Cytiva
2600-100A |
1 to 14 |
23.10 €
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
10052251
|
Cytiva
2611-628 |
1 to 11 |
33.75 €
Each |
In Stock
Log in to see stock. |
|||||
11350674
|
Cytiva
2600-102A |
4 to 7 |
21.20 €
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
11370674
|
Cytiva
2600-104A |
8 to 10 |
21.20 €
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
Used for rapid determination of pH values
- Instant pH readings
- Accurate for wide range of routine pH testing
- Convenient and portable for field use
Specifications
Routine pH testing | |
pH Indicator Dispenser |