Learn More
Cytiva Whatman™ Phenolphthalein Test Paper on Reel
Combines ease-of-use with accuracy and consistency for determination of the neutralization point in weak acid/strong alkali reactions.
Brand: Cytiva 2600-204A
18.76 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Additional Details : Weight : 0.01980kg
Description
Combines ease-of-use with accuracy and consistency for determination of the neutralization point in weak acid/strong alkali reactions.
The convenience of using indicator papers for the rapid determination of pH values has led to many applications in laboratories and industry
- Instant pH readings
- Accurate for a wide range of routine pH testing
- Convenient and portable for field use
- Economical
Specifications
Phenolphthalein | |
For determination of the neutralization point in weak acid/strong alkali reactions | |
Phenolphthalein Test Paper |
8.3 to 10 | |
Each |