missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Polycap™ Disposable Capsules - 36, 75, and 150 SPF

Product Code. 11324614
Change view
Click to view available options
Filtration Area:
1100 cm2
260 cm2
535 cm2
Unit Size:
Each
Pack of 5
Product Code. Filtration Area unitSize
11324614 260 cm2 Each
11354614 535 cm2 Each
11379134 1100 cm2 Pack of 5
3 options
This item is not returnable. View return policy

Product Code. 11324614

Brand: Cytiva 67053600

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides serum prefiltration. Whatman™ Polycap™ Disposable Capsules permit filtration of tissue culture media, nutrients, immunologicals, enzymes, diagnostic standards/controls, biologicals, buffers, virus suspension. Includes inlet vent.

  • Three-layer filter permits filtration of larger volumes
  • Integrity testable
  • With inlet vent
  • Filter Media: Three layers: fine glass microfiber/ultrafine glass microfiber/1.0μm polysulfone

Serum Prefilter Capsules. Retention (with air purge), <2mL

For serum prefiltration. Handle tissue culture media, nutrients, immunologicals, enzymes, diagnostic standards/controls, biologicals, buffers, virus suspensions.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Description Polycap SPF 36 Capsule Filter, 1.0 μm, with SB inlet and outlet (1 pc)
Material (Membrane) Glass Microfiber, Polyethersulfone
Pore Size 1 μm
Filtration Area 260 cm2
For Use With (Application) For serum prefiltration. Handle tissue culture media, nutrients, immunologicals, enzymes, diagnostic standards/controls, biologicals, buffers, virus suspensions.
Housing Material Polypropylene
Max. Pressure 4.1 bar (60 psi)
Outlet Style Stepped Barb
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.