missing translation for 'onlineSavingsMsg'
Learn More

Cytiva™ Whatman™ Polycap Disposable Capsules, 36 AS

Product Code. 10527111
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Inlet Style:
1/2 SB inlet
FNPT
MNPT inlet
SB inlet
Outlet Style:
1/2 SB outlet
FNPT
SB outlet
SB outlet plus filling bell
Sterility:
Non-sterile
Sterile
Unit Size:
Each
Pack of 5
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Inlet Style Outlet Style Sterility Unit Size
10527111 0.2 μm SB inlet SB outlet Sterile Each
11369084 0.2 μm 1/2 SB inlet 1/2 SB outlet Sterile Each
11379084 0.2 μm MNPT inlet SB outlet Sterile Each
11399084 0.45 μm SB inlet SB outlet Sterile Each
11329094 0.2 μm SB inlet SB outlet plus filling bell Sterile Each
11339094 0.2 μm FNPT FNPT Non-sterile Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10527111 Supplier Cytiva™ Supplier No. 67053602
  • ONLINE EXCLUSIVE PRICE
    72.70 €
    67.73 € / Pack of 1
    Save 4.97 €
    6% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Provides highly effective filtration area in a small size. Whatman™ Polycap Disposable AS Capsules, AS 36 filters large volumes and difficult samples with ease.

Recommended for filtering aqueous solutions. Combines a glass microfiber (GMF) prefilter and a nylon membrane, prolonging the life of the filter and allowing larger volumes and difficult samples to be filtered easily.
  • First layer (GMF) acts as a prefilter to ensure longer membrane (0.2, 0.45, and 1.0 μm) life and higher filtration efficiency.
  • Nylon membrane layer is inherently hydrophilic, has low extractables, is biosafe, and has excellent flow rates.
  • Housing thermally fused (no glues, adhesives, or extraneous materials).
  • Integrity-testable by bubble point, pressure decay, or forward flow methods.
  • Provides highly effective filtration area in a small size.
  • Autoclavable; some presterilized with gamma irradiation.
  • Manufactured in clean room facilities under ISO Quality Systems.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Description Polycap AS 36 Capsule Filter, sterile, 0.2 μm, SB inlet and outlet (1 pc)
Material (Membrane) Nylon
Pore Size 0.2 μm
Sterility Sterile
Filtration Area 400 cm2
For Use With (Application) Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.
Housing Material Polypropylene
Inlet Style SB inlet
Max. Pressure 4.1 bar (60 psi)
Outlet Style SB outlet
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.