missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Polydisc Inline Filters
Versatile, cost-effective filters designed for larger volume sample filtration
Brand: Cytiva 6724-5045
Code : 75
Additional Details : Weight : 3.17510kg
Description
Versatile, cost-effective filters designed for larger volume sample filtration
- Filter up to 1L with one device
- Can be used in conjunction with a syringe or connected inline (6-10mm I.D. hose) via stepped hose barbs
- High-purity polypropylene housing maintains sample purity
- 50mm filter devices
- Autoclavable
- Lightweight (11.5g)
- For tissue culture media, reagent preparation, particle counting solutions, pharmaceutical preparations High-throughput polyethersulfone membrane (low protein binding and no surfactants)
- Glass microfiber prefilter extends membrane life and effectively filters heavily contaminated samples
- Sterility cap on outlet (optional)
- No EtO residuals Barbed hose connections fit multiple tubing sizes Integrity testable (bubble point method) Sealed in its own “medical grade” clear blister pack and secured in a protective shelf pack
- Use in vents, inline, exhausts for growth environments, clean or sterile gases, solvents, reagents, drying gases, photoresists Hydrophobic PTFE membrane ideal for chemically aggressive solutions, reagents and organic solvents
- Chemical-resistant housing Integrity testable (bubble point or water breakthrough pressure in situ methods) Biosafe
Specifications
Polydisc AS, PES membrane, 0.45 μm, sterile (10 pcs) | |
0.45 μm | |
20.4 cm2 | |
Polypropylene (PP) | |
Polydisc AS | |
Inline Filter |
PES | |
Sterile | |
Larger volume sample filtration | |
Tubing Nozzle 6 to 10 mm i.d. hose | |
Pk. of 10 | |
50 mm |