missing translation for 'onlineSavingsMsg'
Learn More

Cytiva™ Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.2μm BA83 Protran

Product Code. 15530093
Change view
Click to view available options
Quantity:
1 Each
10/Pk.
5/Pk.
50/Pk.
Format:
Disc
Roll
Sheet
Size (Metric):
140 x 140 mm
15 cm x 3 m
15 x 20 cm
20 x 20 cm
30 cm x 3 m
82 mm dia.
Unit Size:
400g
Each
Pack of 10
Pack of 5
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format Size (Metric) Unit Size
15530093 50/Pk. Sheet 140 x 140 mm Pack of 50
11305304 50/Pk. Disc 82 mm dia. Pack of 50
10589821 1 Each Roll 30 cm x 3 m 400g
11345304 10/Pk. Sheet 15 x 20 cm Pack of 10
11395324 5/Pk. Sheet 20 x 20 cm Pack of 5
11905104 1 Each Roll 15 cm x 3 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15530093 Supplier Cytiva™ Supplier No. 10401403
  • ONLINE EXCLUSIVE PRICE
    629.00 €
    563.79 € / Pack of 50
    Save 65.21 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

100% pure nitrocellulose to ensure the highest binding capacity possible

  • Universal membrane suitable for Western, Southern, and Northern blotting
  • Hydrophilic, so no prewetting with methanol is required
  • High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
  • Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
  • Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
  • Three pore sizes available—0.45μm is ideal for larger molecular weight samples, 0.2μm ensures binding of samples below 20kDa, 0.1μm is used for molecules below 7kDa
  • Also available in circular formats

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 140 mm
Width (Metric) 140 mm
For Use With (Application) Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection
Binding Capacity High
Quantity 50/Pk.
Format Sheet
Material Nitrocellulose
Pore Size 0.2 μm
Size (Metric) 140 x 140 mm
compliance-icons
Product Identifier
  • protran BA 83, 140mm x 140mm
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: nitrocellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.