missing translation for 'onlineSavingsMsg'
Learn More

Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.45μm BA85 Protran Sheets and Rolls

Product Code. 11998905
Change view
Click to view available options
Quantity:
1 Each
Length (Metric):
30 cm
Width (Metric):
3 m
Format:
Roll
Unit Size:
330g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Length (Metric) Width (Metric) Format unitSize
11998905 1 Each 30 cm 3 m Roll 330g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11998905 Supplier Whatman™ Supplier No. 10401196

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

100% pure nitrocellulose to ensure the highest binding capacity possible

  • Universal membrane suitable for Western, Southern, and Northern blotting
  • Hydrophilic, so no prewetting with methanol is required
  • High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
  • Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
  • Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
  • Three pore sizes available—0.45μm is ideal for larger molecular weight samples, 0.2μm ensures binding of samples below 20kDa, 0.1μm is used for molecules below 7kDa
  • Also available in circular formats

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 30 cm
Width (Metric) 3 m
For Use With (Application) Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection
Binding Capacity High
Quantity 1 Each
Format Roll
Material Nitrocellulose
Pore Size 0.45 μm
compliance-icons compliance-icons
Product Identifier
  • ROLLO PROTRAN BA85 0,45µ 3
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: nitrocellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.