missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Qualitative Filter Paper: Grade 1 Circles

Product Code. 11392805
Change view
Click to view available options
Diameter (Metric):
10 mm
15 mm
20 mm
25 mm
30 mm
32 mm
42.5 mm
45 mm
47 mm
55 mm
70 mm
85 mm
90 mm
110 mm
125 mm
150 mm
185 mm
240 mm
270 mm
320 mm
385 mm
400 mm
Quantity:
100/Pk.
400/Pk.
500/Pk.
Unit Size:
Pack of 100
Pack of 400
Pack of 500
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Quantity unitSize
11392805 90 mm 100/Pk. Pack of 100
11350394 15 mm 500/Pk. Pack of 500
11302805 20 mm 400/Pk. Pack of 400
11312805 25 mm 400/Pk. Pack of 400
10193971 30 mm 400/Pk. Pack of 400
11332805 32 mm 100/Pk. Pack of 100
10433891 42.5 mm 100/Pk. Pack of 100
11352805 47 mm 100/Pk. Pack of 100
10292221 55 mm 100/Pk. Pack of 100
11372805 70 mm 100/Pk. Pack of 100
10738611 85 mm 100/Pk. Pack of 100
11302815 110 mm 100/Pk. Pack of 100
10424081 125 mm 100/Pk. Pack of 100
11322815 150 mm 100/Pk. Pack of 100
10434271 185 mm 100/Pk. Pack of 100
11342815 240 mm 100/Pk. Pack of 100
10786262 270 mm 100/Pk. Pack of 100
10134071 320 mm 100/Pk. Pack of 100
15788032 25 mm 100/Pk. Pack of 100
11370394 30 mm 100/Pk. Pack of 100
11340394 385 mm 100/Pk. Pack of 100
11372815 400 mm 100/Pk. Pack of 100
11320394 10 mm 500/Pk. Pack of 500
12494272 45 mm 100/Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
This item is not returnable. View return policy
Product Code. 11392805 Supplier Cytiva Supplier No. 1001090
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Covers a wide range of laboratory applications and is frequently used for clarifying liquids. Whatman™ Qualitative Filter Paper: Grade 1 Circles are widely used standard grade filter papers for routine applications with medium retention and flow rates.

 

These cellulose filters are used in qualitative analytical techniques to determine and identify materials and for clarifying liquids. 
  • Particle Retention: >11μm
  • Porosity: medium
  • Filtration speed: ASTM™, 40 sec.; Herzberg, 150 sec.
  • Surface: smooth
  • Separates precipitates such as lead sulfate, calcium oxalate (hot) and calcium carbonate
  • Used for soil analysis and seed testing procedures in the agricultural industry
  • Used to separate solid foodstuffs from associated liquid or extract liquid in the food industry
  • Atmospheric dust is collected from airflow and the stain-intensity measured 
  • Impregnated with a chromogenic reagent and color formation quantified by optical reflectance for gas detection

 


TRUSTED_SUSTAINABILITY

Specifications

Airflow Rate 13sec./100mL/sq. in.
Basis Weight 87 g/m2
Description Grade 1 Qualitative Filter Paper Standard Grade, circle, 90 mm
Format Disc
Material Cellulose
Particle Retention >11 μm
Filtration Speed 150 sec.
Flow Rate 57mL/min. (water)
For Use With (Application) A widely used standard grade filter paper for routine applications with medium retention and flow rate. It covers a wide range of laboratory applications and is frequently used for clarifying liquids.
Grade Grade 1
Porosity Medium
Quantity 100/Pk.
Thickness (Metric) 180 μm
Type Qualitative Filter Paper
Diameter (English) 3.54 in.
Diameter (Metric) 90 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.