missing translation for 'onlineSavingsMsg'
Learn More

Cytiva™ Whatman™ Sterile Mixed Cellulose Ester Membranes, ME 25

Product Code. 11335314
Change view
Click to view available options
Diameter (Metric):
47 mm
50 mm
Color:
Black
White
Grid Color:
Black
Not Applicable
White
Quantity:
100/Pk.
1000/Pk.
400/Pk.
Unit Size:
Pack of 100
Pack of 1000
Pack of 400
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Color Grid Color Quantity Unit Size
11335314 47 mm White Not Applicable 100/Pk. Pack of 100
11355394 50 mm Black White 100/Pk. Pack of 100
11345394 47 mm Black White 1000/Pk. Pack of 1000
11335394 47 mm Black White 100/Pk. Pack of 100
11345384 47 mm White Black 100/Pk. Pack of 100
13247119 50 mm White Black 400/Pk. Pack of 400
13267119 50 mm White Black 100/Pk. Pack of 100
13237119 47 mm White Black 100/Pk. Pack of 100
13257119 47 mm White Black 400/Pk. Pack of 400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 11335314 Supplier Cytiva™ Supplier No. 10401670
  • ONLINE EXCLUSIVE PRICE
    176.00 €
    165.00 € / Pack of 100
    Save 11.00 €
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cytiva Whatman Sterile Mixed Cellulose Ester Membranes, ME 25, Black, Diameter Outside: 47mm, 1.85 in., Grid Color: White, Autoclavable: Yes, Grade: ME 25, Includes: With interleaving paper and without pad, Membrane Type: WME, Pore Size: 0.45um, Protein Binding Capacity: Medium

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Basis Weight 4.3 to 5.0 mg/cm2
Color White
Material (Membrane) MCE
Pore Size 0.45 μm
Sterility Sterile
For Use With (Application) Microbiological work
Grade ME 25
Grid Color Not Applicable
Includes Interleaving paper
Protein Binding Capacity Medium
Quantity 100/Pk.
Surface Type Plain
Temperature (English) Operating 266°F
Temperature (Metric) Operating 130°C
Type Membrane
Waterflow Rate 12.5 sec./100mL/12.5cm2
Diameter (English) 1.85 in.
Diameter (Metric) 47 mm
Show More Show Less
compliance-icons
Product Identifier
  • ME25 ST, 0.45 µm 47 mm, 100 pack
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.