missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ 47mm Filtration Assembly and Accessories

Product Code. 12431581 Shop All Wheaton Science Products Products
Change view
Click to view available options
Type:
Base
Filtration Assembly
Funnel
Support Screen Gasket
Material:
Borosilicate Glass
Glass
PTFE
PTFE-coated Glass
Unit Size:
Each
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Material unitSize
12431581 Funnel PTFE-coated Glass Each
11718859 Base Glass Each
11764139 Support Screen Gasket PTFE Pack of 6
12411581 Filtration Assembly Borosilicate Glass Each
11561315 Filtration Assembly PTFE-coated Glass Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12431581 Supplier DWK Life Sciences Supplier No. 419374
Add to basket
Add to basket
This item is not returnable. View return policy

Designed for filtering viscous or proteinaceous solutions to produce ultraclean filtrate. Wheaton™ 47mm Filtration Assembly and Accessories are manufactured from the highest quality materials.

  • Includes 300mL graduated funnel
  • Includes PTFE support screen gasket
  • Includes anodized clamp and a No. 8 silicone stopper
  • Includes a 316 stainless steel support with 120 mesh screen
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance USP type I, ASTM E 438 type I, Class A
For Use With (Equipment) Filtration Assembly (47 mm)
Material PTFE-coated Glass
For Use With (Application) Prevents the filter from adhering to the ground glass surfaces
No. per Pack 1 Ea.
Type Funnel
Volume (Metric) 300 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.