missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences 520 Bottle-Top Dispenser Reservoirs

Product Code. 12467191 Shop All Wheaton Science Products Products
Change view
Click to view available options
Volume (Metric):
250 mL
500 mL
1000 mL
2500 mL
Color:
Amber
Clear
Diameter (Metric) Outer Neck:
32 mm
45 mm
Shape:
Round
Square
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color Diameter (Metric) Outer Neck Shape Unit Size
12467191 2500 mL Amber 45 mm Round Each
12457191 1000 mL Amber 45 mm Square Each
12427191 250 mL Amber 32 mm Square Each
12437191 500 mL Clear 32 mm Round Each
12447191 500 mL Amber 32 mm Square Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12467191 Supplier DWK Life Sciences Supplier No. 844030
  • ONLINE EXCLUSIVE PRICE
    72.45 €
    65.92 € / Pack of 1
    Save 6.53 €
    9% Off
Estimated Shipment: 24-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Large selection of glass and Polyethylene reagent bottles, each supplied with one screw cap. Perfectly adapted with Calibrex™ bottle top dispensers.

  • Corresponding neck adapters supplied with the dispensers.

Specifications

Product Type Reagent Bottle
Color Amber
Material Soda Lime Glass
Volume (Metric) 2500 mL
For Use With (Equipment) 520 model bottletop dispenser
Shape Round
Volume (English) 84.53 oz.
Diameter (English) Outer Neck 1.77 in.
Diameter (Metric) Outer Neck 45 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.