missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ 1000mL Filter Flask with 40/35 Joint
Recommended for general filtration
Brand: DWK Life Sciences 635527
290.70 EUR valid until 2024-12-31
Use promo code "21903" to get your promotional price.
Additional Details : Weight : 0.90720kg
Specifications
90mm filtration assembly | |
40 to 60 μm | |
1 Case | |
1 L |
Borosilicate Glass | |
Analysis of particulate or microbiological contamination | |
Safety Coated Filtering Flask |