missing translation for 'onlineSavingsMsg'
Learn More

DWK LIFE SCIENCES™ WHEATON™ Aluminum Open Top Seals, Unlined

Product Code. 10270322 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Natural
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10270322 Natural Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10270322 Supplier DWK LIFE SCIENCES™ Supplier No. 22417801
  • ONLINE EXCLUSIVE PRICE
    135.00 €
    122.98 € / Pack of 1000
    Save 12.02 €
    8% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

WHEATON™ Aluminum Open Top Seals, Unlined expose the stopper for easy sample retrieval with a syringe. Available in a wide range of sizes and colors including natural, red, blue, and green, with good barrier properties and chemical resistance.

Open top seals expose stopper for sample retrieval with a syringe. Wheaton™ Aluminum Open Top Seals are available in a wide range of sizes and colors.

  • Open top design exposes stopper for sample retrieval with a syringe
  • Wide range of styles and sizes available
  • Color selection includes natural, red, blue, and green
  • Good barrier properties
  • Chemically resistant to alcohol and acetone, among others

Specifications

Color Natural
Diameter (Metric) Outer 20 mm
Material Aluminum
Pre-slit No
Sterility Non-sterile
Septum No
Quantity 1000/Cs.
Closure Size 20 mm
For Use With (Equipment) 20 mm Crimp Top Vials
Autoclavable Not Autoclavable
Product Type Open Top Crimp Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.