missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ BOD Bottles

Product Code. 12731638 Shop All Wheaton Science Products Products
Change view
Click to view available options
Volume (Metric):
60 mL
300 mL
No. Sequence:
1 to 24
1 to 36
121 to 144
145 to 168
169 to 192
193 to 216
217 to 240
241 to 264
25 to 48
265 to 288
289 to 312
313 to 336
337 to 360
361 to 384
37 to 72
385 to 408
409 to 432
49 to 72
73 to 96
97 to 120
Specials
Unnumbered
Stopper:
Pennyhead
Robotic
Barcode:
No
Yes
Unit Size:
Pack of 24
Pack of 36
36 product options available for selection
Product selection table with 36 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) No. Sequence Stopper Barcode Unit Size
12731638 300 mL 25 to 48 Robotic Yes Pack of 24
12473711 60 mL Specials Robotic Yes Pack of 36
12463741 300 mL 289 to 312 Pennyhead Yes Pack of 24
11718879 300 mL 49 to 72 Pennyhead Yes Pack of 24
10678714 60 mL Unnumbered Robotic No Pack of 36
12433711 60 mL 37 to 72 Pennyhead Yes Pack of 36
11764179 60 mL 1 to 36 Robotic Yes Pack of 36
11728879 300 mL 73 to 96 Pennyhead Yes Pack of 24
11678308 300 mL Unnumbered Robotic Yes Pack of 24
10590463 300 mL 1 to 24 Robotic Yes Pack of 24
12453721 300 mL 49 to 72 Robotic Yes Pack of 24
12473721 300 mL 73 to 96 Robotic Yes Pack of 24
12413731 300 mL 121 to 144 Robotic Yes Pack of 24
12493721 300 mL 97 to 120 Robotic Yes Pack of 24
12433731 300 mL 145 to 168 Robotic Yes Pack of 24
12453731 300 mL 169 to 192 Robotic Yes Pack of 24
12473731 300 mL 193 to 216 Robotic Yes Pack of 24
12493731 300 mL 217 to 240 Robotic Yes Pack of 24
12413741 300 mL 241 to 264 Robotic Yes Pack of 24
12453741 300 mL 289 to 312 Robotic Yes Pack of 24
12433741 300 mL 265 to 288 Robotic Yes Pack of 24
12473741 300 mL 313 to 336 Robotic Yes Pack of 24
12493741 300 mL 337 to 360 Robotic Yes Pack of 24
12413751 300 mL 361 to 384 Robotic Yes Pack of 24
12433751 300 mL 385 to 408 Robotic Yes Pack of 24
12453751 300 mL 409 to 432 Robotic Yes Pack of 24
11734189 300 mL 1 to 24 Pennyhead Yes Pack of 24
12403721 300 mL Unnumbered Pennyhead Yes Pack of 24
11764219 300 mL 25 to 48 Pennyhead Yes Pack of 24
12403731 300 mL 97 to 120 Pennyhead Yes Pack of 24
12423731 300 mL 121 to 144 Pennyhead Yes Pack of 24
12443731 300 mL 145 to 168 Pennyhead Yes Pack of 24
12463731 300 mL 169 to 192 Pennyhead Yes Pack of 24
12483731 300 mL 193 to 216 Pennyhead Yes Pack of 24
12403741 300 mL 217 to 240 Pennyhead Yes Pack of 24
12423741 300 mL 241 to 264 Pennyhead Yes Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 36 options available.
36 options
This item is not returnable. View return policy
Product Code. 12731638 Supplier DWK Life Sciences Supplier No. 22749702
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Available with either glass robotic or pennyhead stoppers

Bottles

  • Borosilicate glass, 60 or 300mL, numbered or unnumbered
  • With permanent marking areas; numbered versions are also bar-coded
  • Specially designed shoulder radius sweeps all air from inside the bottle during filling
  • 300mL bottles are recommended for the five-day BOD test referenced in Standard Methods for the Examination of Water and Wastewater, 18th Edition, 5210B, 1992
Stoppers
  • Interchangeable borosilicate glass stoppers have tapered bottoms that also prevent air entrapment
  • Flared mouth of bottle forms a water seal around stoppers that prevents air from being drawn in during incubation
  • Stopper joint is compatible with the probes of the leading BOD and DO meters
  • Glass robotic stoppers are compatible with most robotic systems

Use to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action.

Order Info

Dimensions: 300mL bottles are 2.75 diameter x 6.5 in. high (69 x 165mm); 60mL bottles are 1.75 diameter x 4.5 in. high (43 x 115mm)

Specifications

Barcode Yes
Volume (Metric) 300 mL
For Use With (Application) Used to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action
Marking Spot Yes
Material Borosilicate Glass
No. Sequence 25 to 48
Stopper Robotic
Volume (English) 10.14 oz.
Height (English) 6.49 in.
Height (Metric) 16.5 cm
Diameter (English) 2.7 in.
Diameter (Metric) 6.9 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.