Learn More
DWK Life Sciences Wheaton™ Borosilicate Glass Coliwasa Samplers PTFE Seals
Product Code. 12407311 Shop All Wheaton Science Products Products
Change view
Click to view available options
Material:
PTFE
Unit Size:
Pack of 10
Product Code. Material unitSize
12407311 PTFE Pack of 10
1 options
Product Code. 12407311 Supplier DWK Life Sciences Supplier No. 885232
Estimated Shipment: 20-10-2026
Add to basket
Add to basket
Used to collect composite liquid samples from top to bottom in drums, tanks and reservoirs
Positive PTFE seal for borosilicate glass coliwasa.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Used to collect composite liquid samples from top to bottom in drums, tanks and reservoirs
For Use With (Equipment) For Borosilicate Glass coliwasa
Length (English) 42 in.
Length (Metric) 106.5 cm
Material PTFE
Type Seal